Assay Details
Target Gene Details
| Entrez Gene ID: | 3689 | 
| Gene Name: | integrin subunit beta 2 | 
| Gene Aliases: | CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7 | 
| Location: | Chr.21:44885949-44928873 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ITGB2 | NM_001127491.2 | NP_001120963.2 | ||
| AK095992.1 | ||||
| AU279404.1 | ||||
| DA200537.1 | 
Target Gene Details
| Entrez Gene ID: | 100505746 | 
| Gene Name: | ITGB2 antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.21:44921035-44929680 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ITGB2-AS1 | NR_038311.1 | |||
| NR_038312.1 | ||||
| NR_038313.1 | ||||
| NR_038314.1 | ||||
| NR_038315.1 | ||||
| NR_038316.1 | ||||
| DA378472.1 | ||||
| DA942354.1 | ||||
| DA944688.1 | ||||
| DA948979.1 | ||||
| DB115909.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3575398 | Chr.21:44921339 - 44924818 on Build GRCh38 | Gain |  ITGB2-AS1  ITGB2 | 
| esv3647138 | Chr.21:44916487 - 44942284 on Build GRCh38 | Loss |  ITGB2-AS1  ITGB2  FAM207A  LINC01547 | 
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |  ITGB2-AS1  ADARB1  ITGB2  LOC105372836  SSR4P1  PICSAR  LINC00163  FAM207A  LINC01547 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top