Assay Details
Target Gene Details
Entrez Gene ID: | 5316 |
Gene Name: | PBX/knotted 1 homeobox 1 |
Gene Aliases: |
PREP1, pkonx1c |
Location: |
Chr.21:42974510-43033931 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKNOX1 | NM_001286258.1 | NP_001273187.1 | ||
| NM_001320694.1 | NP_001307623.1 | |||
| NM_004571.4 | NP_004562.2 | |||
| AK294794.1 | ||||
| AY142115.1 | AAN34940.1 | |||
| AY196965.1 | AAO45825.1 | |||
| BC000735.2 | AAH00735.1 | |||
| BP369805.1 | ||||
| DR000771.1 | ||||
| Y13613.1 | CAA73934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587660 | Chr.21:42972160 - 43002186 on Build GRCh38 | Loss |
|
| nsv587657 | Chr.21:42752699 - 43087910 on Build GRCh38 | Gain |
|
| esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion |
|
| nsv834105 | Chr.21:42873719 - 43030219 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map