Assay Details
Target Gene Details
Entrez Gene ID: | 104 |
Gene Name: | adenosine deaminase, RNA specific B1 |
Gene Aliases: |
ADAR2, DRABA2, DRADA2, RED1 |
Location: |
Chr.21:45074578-45226563 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADARB1 | NM_001112.3 | NP_001103.1 | ||
| NM_001160230.1 | NP_001153702.1 | |||
| NM_015833.3 | NP_056648.1 | |||
| NM_015834.3 | NP_056649.1 | |||
| NR_027672.1 | ||||
| NR_027673.1 | ||||
| NR_027674.1 | ||||
| NR_073200.1 | ||||
| XM_006723954.2 | XP_006724017.1 | |||
| XM_011529432.1 | XP_011527734.1 | |||
| XM_017028251.1 | XP_016883740.1 | |||
| XM_017028256.1 | XP_016883745.1 | |||
| AB194369.1 | BAE16325.1 | |||
| AB194370.1 | BAE16326.1 | |||
| AB194371.1 | BAE16327.1 | |||
| AB194372.2 | BAE16328.1 | |||
| AB194373.1 | BAE16329.1 | |||
| AF525422.1 | AAM83100.1 | |||
| AF533142.1 | AAM97654.1 | |||
| AY082339.1 | AAL99252.1 | |||
| AY135659.1 | AAN10291.1 | |||
| BC065545.1 | AAH65545.1 | |||
| U76420.1 | AAC51240.1 | |||
| U76421.1 | AAC51241.1 | |||
| U76422.1 | AAC51242.1 | |||
| U82120.1 | AAB61686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|
| nsv3541 | Chr.21:45076500 - 45116714 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map