Assay Details
Target Gene Details
Entrez Gene ID: | 100505746 |
Gene Name: | ITGB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.21:44921035-44929680 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB2-AS1 | NR_038311.1 | 5 | 2101 | |
| NR_038312.1 | 4 | 2058 | ||
| NR_038313.1 | 5 | 2077 | ||
| NR_038314.1 | 4 | 2006 | ||
| NR_038315.1 | 4 | 2002 | ||
| NR_038316.1 | 3 | 1963 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647138 | Chr.21:44916487 - 44942284 on Build GRCh38 | Loss |
|
| nsv834110 | Chr.21:44896268 - 45085236 on Build GRCh38 | Loss |
|

Genomic Map