Assay Details
Target Gene Details
Entrez Gene ID: | 23544 |
Gene Name: | seizure related 6 homolog like |
Gene Aliases: |
- |
Location: |
Chr.22:26169474-26383597 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEZ6L | NM_001184773.1 | NP_001171702.1 | ||
| NM_001184774.1 | NP_001171703.1 | |||
| NM_001184775.1 | NP_001171704.1 | |||
| NM_001184776.1 | NP_001171705.1 | |||
| NM_001184777.1 | NP_001171706.1 | |||
| NM_021115.4 | NP_066938.2 | |||
| XM_005261439.3 | XP_005261496.1 | |||
| XM_005261440.3 | XP_005261497.1 | |||
| XM_006724195.3 | XP_006724258.1 | |||
| XM_011530037.2 | XP_011528339.1 | |||
| XM_011530038.2 | XP_011528340.1 | |||
| XM_011530039.2 | XP_011528341.1 | |||
| XM_017028699.1 | XP_016884188.1 | |||
| XM_017028700.1 | XP_016884189.1 | |||
| AB023144.2 | ||||
| AB041736.1 | BAB40970.1 | |||
| AY358405.1 | AAQ88771.1 | |||
| BC126115.1 | ||||
| BC143851.1 | ||||
| BC143854.1 | ||||
| BM875686.1 | ||||
| CR456574.1 | CAG30460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469709 | Chr.22:26117172 - 26298328 on Build GRCh38 | Loss |
|
| nsv834165 | Chr.22:26112880 - 26291809 on Build GRCh38 | Gain+Loss |
|
| nsv3582 | Chr.22:26165303 - 26198965 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map