Assay Details
Target Gene Details
Entrez Gene ID: | 553158 |
Gene Name: | PRR5-ARHGAP8 readthrough |
Gene Aliases: |
- |
Location: |
Chr.22:44702198-44862784 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR5-ARHGAP8 | NM_181334.5 | NP_851851.3 | ||
| AF195968.1 | AAK58136.1 | |||
| AF195969.1 | AAK58137.1 | |||
| AL355192.1 | CAB90248.1 | |||
| BC132755.1 | ||||
| BC132757.1 | ||||
| BC144382.1 | ||||
| BX335168.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv589108 | Chr.22:44721714 - 44796741 on Build GRCh38 | Loss |
|
| esv33671 | Chr.22:44740089 - 44751574 on Build GRCh38 | Gain |
|
| nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain |
|
| dgv4586n100 | Chr.22:44719617 - 44799761 on Build GRCh38 | Loss |
|
| esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map