Assay Details
Target Gene Details
Entrez Gene ID: | 55615 |
Gene Name: | proline rich 5 |
Gene Aliases: |
FLJ20185k, PP610, PROTOR-1, PROTOR1 |
Location: |
Chr.22:44668547-44737681 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRR5 | NM_001017528.2 | NP_001017528.1 | ||
NM_001017529.2 | NP_001017529.1 | |||
NM_001017530.1 | NP_001017530.1 | |||
NM_001198721.1 | NP_001185650.1 | |||
NM_015366.3 | NP_056181.2 | |||
AK055848.1 | ||||
AK091884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3650 | Chr.22:44650253 - 44683874 on Build GRCh38 | Insertion |
![]() |
nsv829268 | Chr.22:44640618 - 44785937 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1160777 | Chr.22:44606668 - 44692090 on Build GRCh38 | Duplication |
![]() ![]() |
esv2758842 | Chr.22:44630924 - 44814649 on Build GRCh38 | Loss |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
