Assay Details
Target Gene Details
Entrez Gene ID: | 80833 |
Gene Name: | apolipoprotein L3 |
Gene Aliases: |
APOLIII, CG121, CG12_1, apoL-III |
Location: |
Chr.22:36140323-36166177 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL3 | NM_014349.2 | NP_055164.1 | ||
| NM_030644.1 | NP_085147.1 | |||
| NM_145639.1 | NP_663614.1 | |||
| NM_145640.2 | NP_663615.1 | |||
| NM_145641.2 | NP_663616.1 | |||
| NM_145642.2 | NP_663617.1 | |||
| XM_017028946.1 | XP_016884435.1 | |||
| XM_017028947.1 | XP_016884436.1 | |||
| XM_017028948.1 | XP_016884437.1 | |||
| XM_017028949.1 | XP_016884438.1 | |||
| XM_017028952.1 | XP_016884441.1 | |||
| AF070675.1 | AAC83233.1 | |||
| AF305227.1 | AAK20213.1 | |||
| AK025349.1 | ||||
| AY014902.1 | AAG50344.1 | |||
| AY014903.1 | AAG50345.1 | |||
| AY014904.1 | AAG50346.1 | |||
| AY014905.1 | AAG50347.1 | |||
| AY014906.1 | AAG50348.1 | |||
| AY014907.1 | AAG50349.1 | |||
| BC042918.1 | AAH42918.1 | |||
| BC050596.1 | AAH50596.1 | |||
| BF797347.1 | ||||
| DA878288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8093n54 | Chr.22:36137894 - 36187036 on Build GRCh38 | Loss |
|
| esv2751964 | Chr.22:36137737 - 36183513 on Build GRCh38 | Loss |
|
| dgv4563n100 | Chr.22:36142329 - 36186502 on Build GRCh38 | Loss |
|
| esv3575477 | Chr.22:36152724 - 36170331 on Build GRCh38 | Gain |
|
| nsv521866 | Chr.22:36155110 - 36159746 on Build GRCh38 | Loss |
|
| esv3647677 | Chr.22:36137690 - 36164506 on Build GRCh38 | Loss |
|
| nsv526454 | Chr.22:36143804 - 36187036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map