Assay Details
Target Gene Details
Entrez Gene ID: | 492 |
Gene Name: | ATPase plasma membrane Ca2+ transporting 3 |
Gene Aliases: |
CFAP39, CLA2, OPCA, PMCA3, PMCA3a, SCAX1 |
Location: |
Chr.X:153517677-153582929 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2B3 | XM_005274689.3 | XP_005274746.1 | ||
| XM_005274690.3 | XP_005274747.1 | |||
| XM_005274691.3 | XP_005274748.1 | |||
| XM_005274692.3 | XP_005274749.1 | |||
| XM_011531174.2 | XP_011529476.1 | |||
| XM_011531177.2 | XP_011529479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map