Assay Details
Target Gene Details
Entrez Gene ID: | 27330 |
Gene Name: | ribosomal protein S6 kinase A6 |
Gene Aliases: |
PP90RSK4, RSK4 |
Location: |
Chr.X:84058346-84187935 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS6KA6 | NM_014496.4 | 22 | 4641 | NP_055311.1 |
| XM_017029423.1 | 22 | 5811 | XP_016884912.1 | |
| AK023104.1 | 1 | 904 | ||
| AK026301.1 | 1 | 1727 |
More Information
Additional Information:
For this assay, SNP(s) [rs147033792] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map