Assay Details
Target Gene Details
Entrez Gene ID: | 3897 |
Gene Name: | L1 cell adhesion molecule |
Gene Aliases: |
CAML1, CD171, HSAS, HSAS1, MASA, MIC5, N-CAM-L1, N-CAML1, NCAM-L1, S10, SPG1 |
Location: |
Chr.X:153861514-153886174 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L1CAM | NM_001278116.1 | NP_001265045.1 | ||
| DA122729.1 |
Target Gene Details
Entrez Gene ID: | 100996465 |
Gene Name: | lung carcinoma-associated 10 |
Gene Aliases: |
LCAP |
Location: |
Chr.X:153880672-153888990 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCA10 | NR_130768.1 | |||
| AF514420.1 | AAQ08117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map