Assay Details
Target Gene Details
Entrez Gene ID: | 5475 |
Gene Name: | protein phosphatase with EF-hand domain 1 |
Gene Aliases: |
PP7, PPEF, PPP7C, PPP7CA |
Location: |
Chr.X:18675909-18827921 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPEF1 | XM_005274553.2 | XP_005274610.1 | ||
| XM_017029612.1 | XP_016885101.1 |
Target Gene Details
Entrez Gene ID: | 100874004 |
Gene Name: | PPEF1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:18688643-18692687 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPEF1-AS1 | NR_046642.1 | |||
| BM683329.1 | ||||
| BM929366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33517 | Chr.X:18661048 - 19128069 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map