Assay Details
Target Gene Details
Entrez Gene ID: | 6535 |
Gene Name: | solute carrier family 6 member 8 |
Gene Aliases: |
CCDS1, CRT, CRTR, CT1, CTR5 |
Location: |
Chr.X:153688297-153696593 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A8 | NM_001142805.1 | NP_001136277.1 | ||
| NM_001142806.1 | NP_001136278.1 | |||
| NM_005629.3 | NP_005620.1 | |||
| AB209704.1 | BAD92941.1 | |||
| AK295495.1 | ||||
| BC012355.1 | AAH12355.1 | |||
| BC081558.1 | AAH81558.1 | |||
| KJ534942.1 | ||||
| KJ534943.1 | ||||
| KJ534944.1 | ||||
| KJ534945.1 | ||||
| L31409.1 | AAC41688.1 | |||
| S74039.1 | AAB32284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1003299 | Chr.X:153685148 - 153714589 on Build GRCh38 | Gain |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map