Assay Details
Target Gene Details
Entrez Gene ID: | 203522 |
Gene Name: | integrator complex subunit 6 like |
Gene Aliases: |
DDX26B |
Location: |
Chr.X:135520630-135582535 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS6L | NM_182540.4 | NP_872346.3 | ||
| XM_005262394.2 | XP_005262451.1 | |||
| XM_006724740.2 | XP_006724803.1 | |||
| XM_006724741.3 | XP_006724804.1 | |||
| XM_011531311.2 | XP_011529613.1 | |||
| XM_017029342.1 | XP_016884831.1 | |||
| XM_017029343.1 | XP_016884832.1 | |||
| XM_017029344.1 | XP_016884833.1 | |||
| XM_017029345.1 | XP_016884834.1 | |||
| XM_017029346.1 | XP_016884835.1 | |||
| XM_017029347.1 | XP_016884836.1 | |||
| XM_017029348.1 | XP_016884837.1 | |||
| XM_017029349.1 | XP_016884838.1 | |||
| AK096544.1 | BAC04813.1 | |||
| AK308423.1 | ||||
| AL833319.1 | ||||
| BC140715.1 | ||||
| BX648113.1 | ||||
| CN343659.1 | ||||
| CR936620.1 | CAI56763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758895 | Chr.X:135468612 - 136072582 on Build GRCh38 | Gain+Loss |
|
| dgv531n21 | Chr.X:135533826 - 135691851 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map