Assay Details
Target Gene Details
Entrez Gene ID: | 9086 |
Gene Name: | eukaryotic translation initiation factor 1A, Y-linked |
Gene Aliases: |
eIF-4C |
Location: |
Chr.Y:20575711-20593154 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1AY | NM_001278612.1 | NP_001265541.1 | ||
| NM_004681.3 | NP_004672.2 | |||
| AF000987.1 | AAC51834.1 | |||
| AK127664.1 | 1 | 1939 | ||
| BC005248.1 | AAH05248.1 | |||
| BG499107.1 | ||||
| BP331805.1 | ||||
| BT007201.1 | AAP35865.1 | |||
| DQ786267.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10025 | Chr.Y:18865177 - 20859250 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map