Assay Details
Target Gene Details
Entrez Gene ID: | 7042 |
Gene Name: | transforming growth factor beta 2 |
Gene Aliases: |
LDS4, TGF-beta2 |
Location: |
Chr.1:218345334-218444619 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB2 | NM_001135599.2 | 1 | 256 | NP_001129071.1 |
| NM_003238.3 | 1 | 256 | NP_003229.1 | |
| AB209842.1 | 1 | 184 | BAD93079.1 |
Target Gene Details
Entrez Gene ID: | 728463 |
Gene Name: | TGFB2 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.1:218344196-218345678 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB2-AS1 | NR_046268.1 | |||
| BX099841.1 |
More Information
Set Membership: |
|

Genomic Map