Assay Details
Target Gene Details
Entrez Gene ID: | 23095 |
Gene Name: | kinesin family member 1B |
Gene Aliases: |
CMT2, CMT2A, CMT2A1, HMSNII, KLP, NBLST1 |
Location: |
Chr.1:10210706-10381603 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF1B | NM_015074.3 | NP_055889.2 | ||
| NM_183416.3 | NP_904325.2 | |||
| AB011163.2 | BAA25517.2 | |||
| AB040881.2 | 1 | 41 | ||
| AF257176.1 | AAK49332.1 | |||
| AK308121.1 | ||||
| AY139835.1 | AAN17742.1 | |||
| BF940695.1 | ||||
| DA774937.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758920 | Chr.1:10178012 - 10522497 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map