Assay Details
Target Gene Details
Entrez Gene ID: | 6018 |
Gene Name: | rearranged L-myc fusion |
Gene Aliases: |
ZN-15L, ZNF292L |
Location: |
Chr.1:40161369-40240921 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RLF | NM_012421.3 | NP_036553.2 | ||
XM_017002016.1 | XP_016857505.1 | |||
XM_017002017.1 | XP_016857506.1 | |||
XM_017002018.1 | XP_016857507.1 | |||
AK307662.1 | ||||
BC113666.1 | ||||
U22377.1 | AAC50396.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv829660 | Chr.1:40073032 - 40266408 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() |
nsv521874 | Chr.1:40170715 - 40222507 on Build GRCh38 | Loss |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
