Assay Details
Target Gene Details
Entrez Gene ID: | 55083 |
Gene Name: | kinesin family member 26B |
Gene Aliases: |
- |
Location: |
Chr.1:245154985-245703126 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIF26B | NM_018012.3 | NP_060482.2 | ||
| AK125187.1 | BAC86076.1 | |||
| AK293964.1 | ||||
| AK297303.1 | ||||
| AY923834.1 | AAY17361.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891315 | Chr.1:245164783 - 245720849 on Build GRCh38 | Loss |
|
| esv3589431 | Chr.1:245170587 - 245171753 on Build GRCh38 | Loss |
|
| nsv549494 | Chr.1:244991842 - 245263357 on Build GRCh38 | Gain |
|
| nsv524642 | Chr.1:245152917 - 245173470 on Build GRCh38 | Loss |
|
| dgv602n100 | Chr.1:244936197 - 245539982 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map