Assay Details
Target Gene Details
Entrez Gene ID: | 100507564 | 
Gene Name: | uncharacterized LOC100507564 | 
Gene Aliases: | 
	
	- | 
Location: | 
		
			Chr.1:53238610-53242783 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC100507564 | NR_038953.1 | 
Target Copy Number Variation Details
	    DGV Version: |  
	    Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation  |  
		            Location | CNV Subtype  |  
		            Genes | 
|---|---|---|---|
| nsv1011750 | Chr.1:53226981 - 53261502 on Build GRCh38 | Gain | 
							
								 | 
					
| nsv829871 | Chr.1:53200871 - 53383048 on Build GRCh38 | Loss | 
							
								 | 
					
| esv2422213 | Chr.1:53179891 - 53258767 on Build GRCh38 | Duplication | 
							
								 | 
					
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain | 
							
								 | 
					

Genomic Map