Assay Details
Target Gene Details
Entrez Gene ID: | 344148 |
Gene Name: | NCK associated protein 5 |
Gene Aliases: |
ERIH1, ERIH2, NAP5 |
Location: |
Chr.2:132671788-133675181 on Build GRCh38 |
Assay Gene Location: | Within Intron 29 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv583158 | Chr.2:132558519 - 133104527 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv583152 | Chr.2:132369502 - 133631550 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1011421 | Chr.2:132137668 - 132924612 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
