Assay Details
Target Gene Details
Entrez Gene ID: | 6364 |
Gene Name: | C-C motif chemokine ligand 20 |
Gene Aliases: |
CKb4, Exodus, LARC, MIP-3-alpha, MIP-3a, MIP3A, SCYA20, ST38 |
Location: |
Chr.2:227813842-227817564 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCL20 | NM_001130046.1 | NP_001123518.1 | ||
NM_004591.2 | NP_004582.1 | |||
BC020698.1 | AAH20698.1 | |||
D86955.1 | BAA13191.1 | |||
U64197.1 | AAB61534.1 | |||
U77035.1 | AAC50943.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv10221 | Chr.2:227715193 - 228041264 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv834562 | Chr.2:227752511 - 227939244 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
