Assay Details
Target Gene Details
Entrez Gene ID: | 55777 |
Gene Name: | methyl-CpG binding domain protein 5 |
Gene Aliases: |
MRD1 |
Location: |
Chr.2:148021011-148516923 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBD5 | NM_018328.4 | NP_060798.2 | ||
| XM_011511470.2 | XP_011509772.1 | |||
| XM_011511472.2 | XP_011509774.1 | |||
| DA240785.1 | ||||
| EF542797.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015003 | Chr.2:147988165 - 148082084 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map