Assay Details
Target Gene Details
Entrez Gene ID: | 130106 |
Gene Name: | calcium and integrin binding family member 4 |
Gene Aliases: |
KIP4 |
Location: |
Chr.2:26581202-26641798 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CIB4 | NM_001029881.2 | NP_001025052.1 | ||
XM_011532514.2 | XP_011530816.1 | |||
XM_011532517.2 | XP_011530819.1 | |||
XM_017003329.1 | XP_016858818.1 | |||
XM_017003330.1 | XP_016858819.1 | |||
BC127684.1 | ||||
BC127685.1 | ||||
BC140923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv517800 | Chr.2:26449146 - 26617624 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv33316 | Chr.2:25440017 - 26672800 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
