Assay Details
Target Gene Details
Entrez Gene ID: | 57683 |
Gene Name: | zinc finger DBF-type containing 2 |
Gene Aliases: |
- |
Location: |
Chr.2:206274641-206314438 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZDBF2 | NM_001285549.1 | NP_001272478.1 | ||
NM_020923.2 | NP_065974.1 | |||
XM_005246711.3 | XP_005246768.1 | |||
XM_005246712.4 | XP_005246769.1 | |||
XM_005246713.3 | XP_005246770.1 | |||
XM_006712656.3 | XP_006712719.1 | |||
XM_011511532.2 | XP_011509834.1 | |||
XM_011511533.2 | XP_011509835.1 | |||
XM_011511534.2 | XP_011509836.1 | |||
XM_011511535.2 | XP_011509837.1 | |||
XM_017004573.1 | XP_016860062.1 | |||
XM_017004574.1 | XP_016860063.1 | |||
XM_017004575.1 | XP_016860064.1 | |||
AB775780.1 | ||||
AB775781.1 | ||||
AK127271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834516 | Chr.2:206282093 - 206440702 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
