Assay Details
Target Gene Details
Entrez Gene ID: | 214 |
Gene Name: | activated leukocyte cell adhesion molecule |
Gene Aliases: |
CD166, MEMD |
Location: |
Chr.3:105366713-105576913 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ALCAM | NM_001243280.1 | NP_001230209.1 | ||
NM_001243281.1 | NP_001230210.1 | |||
NM_001243283.1 | NP_001230212.1 | |||
NM_001627.3 | NP_001618.2 | |||
AK054632.1 | ||||
AK300362.1 | ||||
AK316113.1 | ||||
AL833702.1 | ||||
AY644765.1 | AAV28819.1 | |||
BC057809.1 | AAH57809.1 | |||
BC137096.1 | ||||
BC137097.1 | ||||
DQ486139.1 | ||||
DQ486140.1 | ||||
L38608.1 | AAB59499.1 | |||
Y10183.1 | CAA71256.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525071 | Chr.3:105286009 - 105690477 on Build GRCh38 | Gain |
![]() ![]() |
nsv829650 | Chr.3:105352049 - 105536192 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
