Assay Details
Target Gene Details
Entrez Gene ID: | 285220 |
Gene Name: | EPH receptor A6 |
Gene Aliases: |
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066 |
Location: |
Chr.3:96814581-97761532 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA6 | NM_001080448.2 | NP_001073917.2 | ||
| NM_001278301.1 | NP_001265230.1 | |||
| XM_006713592.3 | XP_006713655.1 | |||
| XM_011512705.2 | XP_011511007.1 | |||
| XM_011512706.2 | XP_011511008.1 | |||
| XM_011512707.2 | XP_011511009.1 | |||
| XM_017006210.1 | XP_016861699.1 | |||
| XM_017006211.1 | XP_016861700.1 | |||
| XM_017006214.1 | XP_016861703.1 | |||
| XM_017006219.1 | XP_016861708.1 | |||
| AK092565.1 | ||||
| AY358738.1 | AAQ89098.1 | |||
| DA385338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999900 | Chr.3:96819697 - 97180863 on Build GRCh38 | Loss |
|
| nsv520553 | Chr.3:95905623 - 98010484 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map