Assay Details
Target Gene Details
| Entrez Gene ID: | 101927174 | 
| Gene Name: | uncharacterized LOC101927174 | 
| Gene Aliases: | - | 
| Location: | Chr.3:112157-197407 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| LOC101927174 | XM_017007606.1 | XP_016863095.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv1008449 | Chr.3:105120 - 189764 on Build GRCh38 | Gain |  LOC101927174 | 
| esv2759122 | Chr.3:29482 - 756820 on Build GRCh38 | Gain+Loss |  LINC01266  CHL1-AS1  LOC101927174  CHL1 | 
| nsv10236 | Chr.3:132029 - 141096 on Build GRCh38 | Gain+Loss |  LOC101927174 | 
| esv3594872 | Chr.3:36658 - 319672 on Build GRCh38 | Loss |  LOC101927174  CHL1 | 
| dgv8142n54 | Chr.3:21733 - 143598 on Build GRCh38 | Loss |  LOC101927174  LOC102723448 | 
| esv3594883 | Chr.3:123413 - 148333 on Build GRCh38 | Gain |  LOC101927174 | 
| esv2667629 | Chr.3:39576 - 1039187 on Build GRCh38 | Deletion |  LINC01266  CHL1-AS1  CNTN6  LOC101927174  CHL1 | 
| nsv428412 | Chr.3:29467 - 326726 on Build GRCh38 | Gain |  LOC101927174  CHL1 | 
| esv3568528 | Chr.3:129115 - 138468 on Build GRCh38 | Loss |  LOC101927174 | 
| dgv8144n54 | Chr.3:72450 - 170062 on Build GRCh38 | Gain |  LOC101927174 | 
| nsv513028 | Chr.3:133546 - 136894 on Build GRCh38 | Loss |  LOC101927174 | 
| nsv1009220 | Chr.3:44522 - 182907 on Build GRCh38 | Gain |  LOC101927174 | 
| dgv4606n100 | Chr.3:18655 - 164435 on Build GRCh38 | Gain |  LOC101927174  LOC102723448 | 
| esv3893585 | Chr.3:44574 - 267680 on Build GRCh38 | Gain |  LOC101927174  CHL1 | 
| esv3575523 | Chr.3:20214 - 160705 on Build GRCh38 | Gain |  LOC101927174  LOC102723448 | 
| esv3594879 | Chr.3:65856 - 151558 on Build GRCh38 | Gain |  LOC101927174 | 
| esv3594884 | Chr.3:134250 - 171681 on Build GRCh38 | Loss |  LOC101927174 | 
| dgv4613n100 | Chr.3:52474 - 1799230 on Build GRCh38 | Loss |  LINC01266  CHL1-AS1  CNTN6  LOC101927174  CHL1 | 
| nsv997732 | Chr.3:93308 - 148728 on Build GRCh38 | Gain |  LOC101927174 | 
| esv2752007 | Chr.3:25192 - 175961 on Build GRCh38 | Gain |  LOC101927174 | 
| nsv998580 | Chr.3:111816 - 138468 on Build GRCh38 | Loss |  LOC101927174 | 
| nsv589242 | Chr.3:47730 - 388999 on Build GRCh38 | Gain |  CHL1-AS1  LOC101927174  CHL1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic