Assay Details
Target Gene Details
Entrez Gene ID: | 79740 |
Gene Name: | zinc finger B-box domain containing |
Gene Aliases: |
- |
Location: |
Chr.3:167178399-167380405 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZBBX |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv592495 | Chr.3:167077165 - 167655374 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
dgv4982n100 | Chr.3:167184267 - 167248071 on Build GRCh38 | Gain |
![]() |
