Assay Details
Target Gene Details
Entrez Gene ID: | 116984 |
Gene Name: | ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 2 |
Gene Aliases: |
CENTD1, PARX |
Location: |
Chr.4:36005186-36244832 on Build GRCh38 |
Assay Gene Location: | Within Intron 37 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARAP2 | XM_017007698.1 | XP_016863187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829905 | Chr.4:35895023 - 36085836 on Build GRCh38 | Gain |
|
| nsv829906 | Chr.4:36026233 - 36186441 on Build GRCh38 | Gain |
|
| esv3569373 | Chr.4:35669991 - 36407345 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112556546] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map