Assay Details
Target Gene Details
Entrez Gene ID: | 285550 |
Gene Name: | family with sequence similarity 200 member B |
Gene Aliases: |
- |
Location: |
Chr.4:15681729-15690447 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM200B | NM_001145191.1 | NP_001138663.1 | ||
| XM_017008048.1 | XP_016863537.1 | |||
| DA408954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv315n21 | Chr.4:15609148 - 15690513 on Build GRCh38 | Loss |
|
| nsv949802 | Chr.4:15647778 - 15695177 on Build GRCh38 | Duplication |
|
| nsv428439 | Chr.4:15555338 - 15747897 on Build GRCh38 | Gain+Loss |
|
| esv2759227 | Chr.4:15442993 - 15747897 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79686577] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map