| dgv5499n100 | 
						
							
								Chr.4:187514678 - 188139456
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| esv2752041 | 
						
							
								Chr.4:187118866 - 188100311
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 ZFP42
							
								 TRIML2
							
								 LOC339975
							
						 | 
					
				
		    		
						| nsv1031595 | 
						
							
								Chr.4:187935080 - 188218104
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1031097 | 
						
							
								Chr.4:187304029 - 188382785
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
								 LOC339975
							
						 | 
					
				
		    		
						| dgv9439n54 | 
						
							
								Chr.4:186930134 - 188890842
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LOC101930028
							
								 LINC01060
							
								 TRIML2
							
								 LOC339975
							
						 | 
					
				
		    		
						| esv2763831 | 
						
							
								Chr.4:187562041 - 189231692
							    on Build GRCh38
						
							
						 | 
						Gain+Loss | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LOC101930028
							
								 LINC01060
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv596456 | 
						
							
								Chr.4:187865684 - 188095782
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| dgv9440n54 | 
						
							
								Chr.4:187534043 - 188139952
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv470098 | 
						
							
								Chr.4:187776941 - 188300053
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv10629 | 
						
							
								Chr.4:187724452 - 188143423
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| dgv9441n54 | 
						
							
								Chr.4:187540004 - 188871837
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LOC101930028
							
								 LINC01060
							
								 TRIML2
							
						 | 
					
				
		    		
						| dgv332n21 | 
						
							
								Chr.4:187966266 - 188141060
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| esv32613 | 
						
							
								Chr.4:187056789 - 188514091
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LINC01060
							
								 TRIML2
							
								 LOC339975
							
						 | 
					
				
		    		
						| nsv1030775 | 
						
							
								Chr.4:187983480 - 188060286
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 ZFP42
							
						 | 
					
				
		    		
						| dgv5496n100 | 
						
							
								Chr.4:187167070 - 188594145
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LINC01060
							
								 TRIML2
							
								 LOC339975
							
						 | 
					
				
		    		
						| esv3569882 | 
						
							
								Chr.4:187528921 - 188584766
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LINC01060
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1027385 | 
						
							
								Chr.4:187992714 - 188017314
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 ZFP42
							
						 | 
					
				
		    		
						| nsv830173 | 
						
							
								Chr.4:187978318 - 188202820
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1022128 | 
						
							
								Chr.4:187957299 - 188139456
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 TRIML1
							
								 ZFP42
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1017684 | 
						
							
								Chr.4:187530002 - 188872654
							    on Build GRCh38
						
							
						 | 
						Loss | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LOC101930028
							
								 LINC01060
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1031236 | 
						
							
								Chr.4:187526594 - 189035370
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 LOC101930028
							
								 LINC01060
							
								 TRIML2
							
						 | 
					
				
		    		
						| nsv1022361 | 
						
							
								Chr.4:186346140 - 188438508
							    on Build GRCh38
						
							
						 | 
						Gain | 
						
							
								 LOC100506272
							
								 TRIML1
							
								 ZFP42
							
								 MTNR1A
							
								 TRIML2
							
								 FAT1
							
								 LOC339975
							
								 F11-AS1
							
						 |