Assay Details
Target Gene Details
Entrez Gene ID: | 5229 |
Gene Name: | protein geranylgeranyltransferase type I subunit beta |
Gene Aliases: |
BGGI, GGTI |
Location: |
Chr.5:115200247-115262887 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGGT1B | NM_005023.3 | 9 | 2206 | NP_005014.2 |
| XM_005272020.3 | 10 | 2109 | XP_005272077.2 | |
| XM_011543490.2 | XP_011541792.1 | |||
| XM_011543492.2 | XP_011541794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv980715 | Chr.5:115209314 - 115212445 on Build GRCh38 | Duplication |
|
| nsv830454 | Chr.5:115118167 - 115271462 on Build GRCh38 | Gain+Loss |
|
| nsv1029371 | Chr.5:115205731 - 115255194 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72813817] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map