Assay Details
Target Gene Details
Entrez Gene ID: | 5229 |
Gene Name: | protein geranylgeranyltransferase type I subunit beta |
Gene Aliases: |
BGGI, GGTI |
Location: |
Chr.5:115200247-115262887 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGGT1B | XM_005272020.3 | 10 | 6526 | XP_005272077.2 |
| XM_011543490.2 | XP_011541792.1 | |||
| XM_011543492.2 | XP_011541794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830454 | Chr.5:115118167 - 115271462 on Build GRCh38 | Gain+Loss |
|
| nsv1029371 | Chr.5:115205731 - 115255194 on Build GRCh38 | Loss |
|
| nsv830453 | Chr.5:114941564 - 115210839 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map