Assay Details
Target Gene Details
Entrez Gene ID: | 55914 |
Gene Name: | erbb2 interacting protein |
Gene Aliases: |
ERBB2IP, HEL-S-78, LAP2 |
Location: |
Chr.5:65926554-66081024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERBIN | NM_001006600.2 | NP_001006600.1 | ||
| NM_001253697.1 | NP_001240626.1 | |||
| NM_001253698.1 | NP_001240627.1 | |||
| NM_001253699.1 | NP_001240628.1 | |||
| NM_001253701.1 | NP_001240630.1 | |||
| NM_018695.3 | NP_061165.1 | |||
| XM_005248554.2 | XP_005248611.1 | |||
| XM_005248555.3 | XP_005248612.1 | |||
| XM_006714660.3 | XP_006714723.1 | |||
| XM_017009636.1 | XP_016865125.1 | |||
| XM_017009637.1 | XP_016865126.1 | |||
| AF263744.1 | AAF77048.1 | |||
| AF276423.1 | AAK69431.1 | |||
| AK295708.1 | ||||
| AK304693.1 | ||||
| BC050692.1 | AAH50692.1 | |||
| BC126464.1 | ||||
| BC144075.1 | ||||
| DA723889.1 | ||||
| DR000131.1 | ||||
| EU668330.1 |
Target Gene Details
Entrez Gene ID: | 100303749 |
Gene Name: | LSM3 homolog, U6 small nuclear RNA associated (S. cerevisiae) pseudogene |
Gene Aliases: |
- |
Location: |
Chr.5:65944805-65945573 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100303749 | NR_028474.1 | 1 | 166 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894205 | Chr.5:65896048 - 65955310 on Build GRCh38 | Gain |
|
| nsv830327 | Chr.5:65910646 - 66032995 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map