Assay Details
Target Gene Details
Entrez Gene ID: | 105379198 |
Gene Name: | uncharacterized LOC105379198 |
Gene Aliases: |
- |
Location: |
Chr.5:127218484-127229743 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105379198 | XM_011543790.2 | XP_011542092.1 |
Target Gene Details
Entrez Gene ID: | 84466 |
Gene Name: | multiple EGF like domains 10 |
Gene Aliases: |
EMARDD |
Location: |
Chr.5:127229519-127461222 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEGF10 | XM_017009987.1 | 1 | 143 | XP_016865476.1 |
| AK123568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1033607 | Chr.5:127049489 - 127968767 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map