Assay Details
Target Gene Details
Entrez Gene ID: | 821 |
Gene Name: | calnexin |
Gene Aliases: |
CNX, IP90, P90 |
Location: |
Chr.5:179678631-179731641 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CANX | XM_011534665.2 | XP_011532967.1 |
Target Gene Details
Entrez Gene ID: | 646019 |
Gene Name: | chibby family member 3 |
Gene Aliases: |
- |
Location: |
Chr.5:179677969-179681830 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBY3 | NM_001164444.1 | NP_001157916.1 | ||
| XM_017009726.1 | XP_016865215.1 | |||
| BX110582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv214n111 | Chr.5:179620185 - 179696634 on Build GRCh38 | Duplication |
|
| esv3570595 | Chr.5:179676712 - 179683849 on Build GRCh38 | Loss |
|
| nsv830560 | Chr.5:179612211 - 179748757 on Build GRCh38 | Loss |
|
| nsv5160 | Chr.5:179675417 - 179709494 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map