Assay Details
Target Gene Details
Entrez Gene ID: | 39 |
Gene Name: | acetyl-CoA acetyltransferase 2 |
Gene Aliases: |
- |
Location: |
Chr.6:159761957-159779112 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACAT2 | NM_001303253.1 | NP_001290182.1 | ||
| NM_005891.2 | NP_005882.2 | |||
| AB208993.1 | BAD92230.1 | |||
| AF356877.1 | AAM00223.1 | |||
| AK225089.1 | ||||
| AK225244.1 | ||||
| AK291080.1 | ||||
| AK294273.1 | ||||
| BC000408.2 | AAH00408.1 | |||
| BP210609.1 | ||||
| S70154.1 | AAB30856.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map