Assay Details
Target Gene Details
Entrez Gene ID: | 60561 |
Gene Name: | RAD50 interactor 1 |
Gene Aliases: |
RINT-1 |
Location: |
Chr.7:105532085-105567688 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RINT1 | NM_021930.4 | NP_068749.3 | ||
XM_017012496.1 | XP_016867985.1 | |||
XM_017012497.1 | XP_016867986.1 | |||
XM_017012498.1 | XP_016867987.1 | |||
AF317622.1 | AAG42101.1 | |||
AK055508.1 | ||||
AK057757.1 | ||||
AK308126.1 | ||||
BC068483.1 | AAH68483.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464668 | Chr.7:105448246 - 105568045 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3614417 | Chr.7:105370987 - 105549944 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv2759549 | Chr.7:105117803 - 105568045 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
