Assay Details
Target Gene Details
Entrez Gene ID: | 50652 |
Gene Name: | prostate cancer associated 3 (non-protein coding) |
Gene Aliases: |
DD3, NCRNA00019, PCAT3 |
Location: |
Chr.9:76764436-76787569 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCA3 | NR_015342.2 | |||
| NR_132312.1 | ||||
| NR_132313.1 | ||||
| AF103907.1 | ||||
| AY894120.1 | ||||
| DQ374660.1 | ||||
| DQ374661.1 |
Target Gene Details
Entrez Gene ID: | 158471 |
Gene Name: | prune homolog 2 |
Gene Aliases: |
BMCC1, BNIPXL, C9orf65, KIAA0367 |
Location: |
Chr.9:76611376-76906220 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2738601 | Chr.9:76768646 - 76769346 on Build GRCh38 | Deletion |
|
| esv2759697 | Chr.9:76750949 - 77058991 on Build GRCh38 | Loss |
|
| nsv1040363 | Chr.9:76631450 - 77096821 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79708259] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map