Assay Details
Target Gene Details
Entrez Gene ID: | 1955 |
Gene Name: | multiple EGF like domains 9 |
Gene Aliases: |
EGFL5 |
Location: |
Chr.9:120600918-120714487 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEGF9 | NM_001080497.2 | NP_001073966.2 | ||
| AB011542.1 | BAA32470.1 | |||
| AK295380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33143 | Chr.9:120491423 - 120708811 on Build GRCh38 | Loss |
|
| nsv831711 | Chr.9:120503030 - 120705363 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map