Assay Details
Target Gene Details
Entrez Gene ID: | 102724719 |
Gene Name: | uncharacterized LOC102724719 |
Gene Aliases: |
- |
Location: |
Chr.10:51062579-51068552 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724719 | NR_120678.1 | |||
| BF448553.1 |
Target Gene Details
Entrez Gene ID: | 5592 |
Gene Name: | protein kinase, cGMP-dependent, type I |
Gene Aliases: |
AAT8, PKG, PRKG1B, PRKGR1B, cGK, cGK 1, cGK1, cGKI, cGKI-BETA, cGKI-alpha |
Location: |
Chr.10:50991151-52298350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKG1 | NM_001098512.2 | NP_001091982.1 | ||
| AK300110.1 | ||||
| D45864.1 | BAA08297.1 | |||
| DC310312.1 | ||||
| DC379897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831873 | Chr.10:50950850 - 51139780 on Build GRCh38 | Gain |
|
| nsv1036645 | Chr.10:51045201 - 51399316 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map