Assay Details
Target Gene Details
Entrez Gene ID: | 219736 |
Gene Name: | storkhead box 1 |
Gene Aliases: |
C10orf24 |
Location: |
Chr.10:68827537-68895942 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STOX1 | NM_001130159.2 | NP_001123631.1 | ||
NM_001130160.2 | NP_001123632.1 | |||
NM_001130161.2 | NP_001123633.1 | |||
NM_152709.4 | NP_689922.3 | |||
XM_011539454.2 | XP_011537756.1 | |||
AK124681.1 | BAC85925.1 | |||
AY842014.1 | AAW48524.1 | |||
AY842015.1 | AAW48525.1 | |||
AY842016.1 | AAW48526.1 | |||
AY842017.1 | AAW48527.1 | |||
BC140011.1 | ||||
DA118241.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759764 | Chr.10:68697962 - 68871556 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv1047983 | Chr.10:68414663 - 68849311 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
