Assay Details
Target Gene Details
Entrez Gene ID: | 25906 |
Gene Name: | anaphase promoting complex subunit 15 |
Gene Aliases: |
APC15, C11orf51, HSPC020 |
Location: |
Chr.11:72106254-72112793 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC15 | XM_017017488.1 | XP_016872977.1 | ||
| XM_017017489.1 | XP_016872978.1 | |||
| XM_017017490.1 | XP_016872979.1 | |||
| XM_017017491.1 | XP_016872980.1 | |||
| XM_017017492.1 | XP_016872981.1 | |||
| XM_017017493.1 | XP_016872982.1 | |||
| XM_017017494.1 | XP_016872983.1 | |||
| XM_017017495.1 | XP_016872984.1 | |||
| AK311146.1 |
Target Gene Details
Entrez Gene ID: | 220074 |
Gene Name: | leucine rich transmembrane and O-methyltransferase domain containing |
Gene Aliases: |
CFAP111, DFNB63, LRRC51 |
Location: |
Chr.11:72079982-72110782 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTOMT | NM_001145308.4 | 6 | 1376 | NP_001138780.1 |
| NM_001145309.3 | 8 | 1597 | NP_001138781.1 | |
| NM_001145310.3 | 8 | 1477 | NP_001138782.1 | |
| XM_017017356.1 | 3 | 1991 | XP_016872845.1 | |
| XM_017017357.1 | 2 | 2914 | XP_016872846.1 | |
| XM_017017358.1 | 2 | 2914 | XP_016872847.1 | |
| AF289588.1 | 4 | 459 | AAL55772.1 | |
| AK302772.1 | 6 | 870 | ||
| EU627069.1 | 8 | 1069 | ||
| EU627070.1 | 8 | 949 |
More Information
Set Membership: |
|

Genomic Map