Assay Details
Target Gene Details
Entrez Gene ID: | 26610 |
Gene Name: | elongator acetyltransferase complex subunit 4 |
Gene Aliases: |
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4 |
Location: |
Chr.11:31509729-31784525 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELP4 | NM_001288725.1 | NP_001275654.1 | ||
| NM_001288726.1 | NP_001275655.1 | |||
| NM_019040.4 | NP_061913.3 | |||
| AK000505.1 | BAA91212.1 | |||
| AK304885.1 | ||||
| AL136677.1 | CAB66612.1 | |||
| BC012514.1 | AAH12514.1 | |||
| HY127287.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv467785 | Chr.11:31467343 - 31674788 on Build GRCh38 | Loss |
|
| dgv1740n54 | Chr.11:31433428 - 31674788 on Build GRCh38 | Loss |
|
| esv2751022 | Chr.11:31041408 - 31526385 on Build GRCh38 | Loss |
|
| esv3625830 | Chr.11:31513026 - 31809558 on Build GRCh38 | Gain |
|
| nsv529004 | Chr.11:30992197 - 31700936 on Build GRCh38 | Loss |
|
| nsv1052451 | Chr.11:31495114 - 31533123 on Build GRCh38 | Loss |
|
| nsv553949 | Chr.11:31296288 - 31518040 on Build GRCh38 | Loss |
|
| nsv553988 | Chr.11:31513808 - 31629642 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map