Assay Details
Target Gene Details
Entrez Gene ID: | 54477 |
Gene Name: | pleckstrin homology domain containing A5 |
Gene Aliases: |
PEPP-2, PEPP2 |
Location: |
Chr.12:19129680-19376400 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1041486 | Chr.12:19107917 - 19493057 on Build GRCh38 | Gain |
|
| esv2422261 | Chr.12:19109497 - 19353437 on Build GRCh38 | Duplication |
|
| esv2422206 | Chr.12:18800088 - 19411427 on Build GRCh38 | Duplication |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111762028] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map