Assay Details
Target Gene Details
Entrez Gene ID: | 84530 |
Gene Name: | serine/arginine repetitive matrix 4 |
Gene Aliases: |
KIAA1853, MU-MB-2.76, nSR100 |
Location: |
Chr.12:118981495-119163051 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRRM4 | NM_194286.3 | NP_919262.2 | ||
| AB058756.1 | ||||
| AK291361.1 | ||||
| BC152471.1 | ||||
| BC167162.1 | ||||
| BC167163.1 | ||||
| KC995185.1 | ||||
| KC995186.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438238 | Chr.12:118997948 - 119003407 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map