Assay Details
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRG1 | NM_005810.3 | NP_005801.3 | ||
| XM_017018682.1 | XP_016874171.1 | |||
| XM_017018683.1 | XP_016874172.1 | |||
| XM_017018684.1 | XP_016874173.1 | |||
| XM_017018685.1 | XP_016874174.1 | |||
| AF034952.1 | AAC34731.1 | |||
| AF081675.1 | AAC32200.1 | |||
| AF097358.1 | AAD03719.1 | |||
| AK301367.1 | ||||
| AK316337.1 | ||||
| BC012621.1 | AAH12621.1 | |||
| BQ212545.1 | ||||
| DT217650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv832327 | Chr.12:8929183 - 9093624 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map