Assay Details
Target Gene Details
Entrez Gene ID: | 64786 |
Gene Name: | TBC1 domain family member 15 |
Gene Aliases: |
RAB7-GAP |
Location: |
Chr.12:71839707-71926849 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D15 | NM_001146213.1 | NP_001139685.2 | ||
| NM_001146214.1 | NP_001139686.1 | |||
| NM_022771.4 | NP_073608.4 | |||
| NR_027449.1 | ||||
| XM_006719564.2 | XP_006719627.1 | |||
| XM_006719565.3 | XP_006719628.1 | |||
| XM_006719566.2 | XP_006719629.1 | |||
| XM_006719567.2 | XP_006719630.1 | |||
| XM_011538679.1 | XP_011536981.1 | |||
| XM_011538680.1 | 1 | 66 | XP_011536982.1 | |
| XM_011538681.2 | XP_011536983.1 | |||
| XM_011538682.1 | XP_011536984.1 | |||
| XM_017019824.1 | XP_016875313.1 | |||
| XM_017019825.1 | XP_016875314.1 | |||
| XM_017019826.1 | XP_016875315.1 | |||
| XM_017019828.1 | XP_016875317.1 | |||
| AB449901.1 | ||||
| AK022147.1 | BAB13971.1 | |||
| AK225221.1 | ||||
| AK225747.1 | ||||
| AK292306.1 | ||||
| AK297626.1 | ||||
| AK307922.1 | ||||
| AL833639.1 | ||||
| BC028352.2 | AAH28352.1 | |||
| DA438409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759907 | Chr.12:71708684 - 71914393 on Build GRCh38 | Gain |
|
| nsv559329 | Chr.12:71748682 - 71886804 on Build GRCh38 | Loss |
|
| esv3892218 | Chr.12:71785292 - 71892336 on Build GRCh38 | Gain |
|
| esv35030 | Chr.12:71778953 - 71879053 on Build GRCh38 | Gain |
|
| esv3629916 | Chr.12:71783025 - 71889798 on Build GRCh38 | Gain |
|
| nsv818924 | Chr.12:71788125 - 71886804 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map