Assay Details
Target Gene Details
Entrez Gene ID: | 341392 |
Gene Name: | acyl-CoA synthetase medium-chain family member 4 |
Gene Aliases: |
- |
Location: |
Chr.12:7304332-7328373 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSM4 | NM_001080454.1 | NP_001073923.1 |
Target Gene Details
Entrez Gene ID: | 283316 |
Gene Name: | CD163 molecule like 1 |
Gene Aliases: |
CD163B, M160, SCARI2 |
Location: |
Chr.12:7320510-7444185 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD163L1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv448e212 | Chr.12:7280402 - 7344654 on Build GRCh38 | Gain |
|
| dgv1349n100 | Chr.12:7274380 - 7336757 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map